Merck
  • Reduced calcium levels and accumulation of abnormal insulin granules in stem cell models of HNF1A deficiency.
Reduced calcium levels and accumulation of abnormal insulin granules in stem cell models of HNF1A deficiency.
Communications biology (2022-08-03)
Bryan J González, Haoquan Zhao, Jacqueline Niu, Damian J Williams, Jaeyop Lee, Chris N Goulbourne, Yuan Xing, Yong Wang, Jose Oberholzer, Maria H Blumenkrantz, Xiaojuan Chen, Charles A LeDuc, Wendy K Chung, Henry M Colecraft, Jesper Gromada, Yufeng Shen, Robin S Goland, Rudolph L Leibel, Dieter Egli
ZUSAMMENFASSUNG
Mutations in HNF1A cause Maturity Onset Diabetes of the Young (HNF1A-MODY). To understand mechanisms of β-cell dysfunction, we generated stem cell-derived pancreatic endocrine cells with hypomorphic mutations in HNF1A. HNF1A-deficient β-cells display impaired basal and glucose stimulated-insulin secretion, reduced intracellular calcium levels in association with a reduction in CACNA1A expression, and accumulation of abnormal insulin granules in association with SYT13 down-regulation. Knockout of CACNA1A and SYT13 reproduce the relevant phenotypes. In HNF1A deficient β-cells, glibenclamide, a sulfonylurea drug used in the treatment of HNF1A-MODY patients, increases intracellular calcium, and restores insulin secretion. While insulin secretion defects are constitutive in β-cells null for HNF1A, β-cells heterozygous for hypomorphic HNF1A (R200Q) mutations lose the ability to secrete insulin gradually; this phenotype is prevented by correction of the mutation. Our studies illuminate the molecular basis for the efficacy of treatment of HNF1A-MODY with sulfonylureas, and suggest promise for the use of cell therapies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Tubulin-III in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Glybenclamid, ≥99% (HPLC)
Sigma-Aldrich
γγ-Sekretase-Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.