Merck
  • Comparative Analysis of Policosanols Related to Growth Times from the Seedlings of Various Korean Oat (Avena sativa L.) Cultivars and Screening for Adenosine 5'-Monophosphate-Activated Protein Kinase (AMPK) Activation.
Comparative Analysis of Policosanols Related to Growth Times from the Seedlings of Various Korean Oat (Avena sativa L.) Cultivars and Screening for Adenosine 5'-Monophosphate-Activated Protein Kinase (AMPK) Activation.
Plants (Basel, Switzerland) (2022-07-28)
Han-Gyeol Lee, So-Yeun Woo, Hyung-Jae Ahn, Ji-Yeong Yang, Mi-Ja Lee, Hyun-Young Kim, Seung-Yeob Song, Jin-Hwan Lee, Woo-Duck Seo
ZUSAMMENFASSUNG
The objectives of this research were to evaluate the policosanol profiles and adenosine-5'-monophosphate-activated protein kinase (AMPK) properties in the seedlings of Korean oat (Avena sativa L.) cultivars at different growth times. Nine policosanols in the silylated hexane extracts were detected using GC-MS and their contents showed considerable differences; specifically, hexacosanol (6) exhibited the highest composition, constituting 88-91% of the total average content. Moreover, the average hexacosanol (6) contents showed remarkable variations of 337.8 (5 days) → 416.8 (7 days) → 458.9 (9 days) → 490.0 (11 days) → 479.2 (13 days) → 427.0 mg/100 g (15 days). The seedlings collected at 11 days showed the highest average policosanol content (541.7 mg/100 g), with the lowest content being 383.4 mg/100 g after 5 days. Interestingly, policosanols from oat seedlings grown for 11 days induced the most prevalent phenotype of AMPK activation in HepG2 cells, indicating that policosanols are an excellent AMPK activator.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution