- Whole-mount immunofluorescence staining of mesenchymal progenitors in murine plantaris muscle.
Whole-mount immunofluorescence staining of mesenchymal progenitors in murine plantaris muscle.
STAR protocols (2022-08-10)
Tamaki Kurosawa, Madoka Ikemoto-Uezumi, Akihiro Kaneshige, So-Ichiro Fukada, Akiyoshi Uezumi
PMID35942341
ZUSAMMENFASSUNG
We recently demonstrated that mesenchymal progenitors play a critical role in regulating satellite cell-dependent myonuclear accretion during overload-induced muscle hypertrophy. Here, we describe the detailed protocol for whole-mount immunofluorescence staining of mesenchymal progenitors in mouse plantaris muscle. Z-stack image reconstruction provides a whole-cell image and enables examination of YAP nuclear translocation in mesenchymal progenitors induced by overload. For complete details on the use and execution of this protocol, please refer to Kaneshige et al. (2022a).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%