- High performance PtRuIr catalysts supported on carbon nanotubes for the anodic oxidation of methanol.
High performance PtRuIr catalysts supported on carbon nanotubes for the anodic oxidation of methanol.
Journal of the American Chemical Society (2006-03-16)
Shijun Liao, Kerry-Anne Holmes, Haralampos Tsaprailis, Viola I Birss
PMID16536508
ZUSAMMENFASSUNG
We report the formation of a new PtRuIr catalyst using an organic colloid synthesis method, involving acetone as the solvent, ethylene glycol as the reducing agent, citrate as a complexing agent and stabilizer, and multiwall carbon nanotubes (CNT, diameter 8-10 nm) as the support. This catalyst has a very high real surface area and is highly active toward the oxidation of methanol, relevant to fuel cell applications. The Ir component appears to act as a promoter, and the splitting of the Pt(111) XRD feature into four peaks and the shift to larger d spacing reflect the high dispersion of the metallic components.