Merck
  • Surface-and redox-active multifunctional polyphenol-derived poly (ionic liquid) s: controlled synthesis and characterization
Surface-and redox-active multifunctional polyphenol-derived poly (ionic liquid) s: controlled synthesis and characterization
Macromolecules (2016)
Patil N, et al.