Merck
Stratifin as a novel diagnostic biomarker in serum for diffuse alveolar damage.
Nature communications (2022-10-05)
Noriaki Arakawa, Atsuhito Ushiki, Mitsuhiro Abe, Shinichiro Matsuyama, Yoshinobu Saito, Takeru Kashiwada, Yasushi Horimasu, Akihiko Gemma, Koichiro Tatsumi, Noboru Hattori, Kenji Tsushima, Keiko Miyashita, Kosuke Saito, Ryosuke Nakamura, Takeshi Toyoda, Kumiko Ogawa, Motonobu Sato, Kazuhiko Takamatsu, Kazuhiko Mori, Takayoshi Nishiya, Takashi Izumi, Yasuo Ohno, Yoshiro Saito, Masayuki Hanaoka
ZUSAMMENFASSUNG
Among the various histopathological patterns of drug-induced interstitial lung disease (DILD), diffuse alveolar damage (DAD) is associated with poor prognosis. However, there is no reliable biomarker for its accurate diagnosis. Here, we show stratifin/14-3-3σ (SFN) as a biomarker candidate found in a proteomic analysis. The study includes two independent cohorts (including totally 26 patients with DAD) and controls (total 432 samples). SFN is specifically elevated in DILD patients with DAD, and is superior to the known biomarkers, KL-6 and SP-D, in discrimination of DILD patients with DAD from patients with other DILD patterns or other lung diseases. SFN is also increased in serum from patients with idiopathic DAD, and in lung tissues and bronchoalveolar lavage fluid of patients with DAD. In vitro analysis using cultured lung epithelial cells suggests that extracellular release of SFN occurs via p53-dependent apoptosis. We conclude that serum SFN is a promising biomarker for DAD diagnosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SFN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-14-3-3σ Antibody, clone CS112-2A8, clone CS112-2A8, Upstate®, from mouse