Merck
  • The interferon-inducible protein viperin controls cancer metabolic reprogramming to enhance cancer progression.
The interferon-inducible protein viperin controls cancer metabolic reprogramming to enhance cancer progression.
The Journal of clinical investigation (2022-10-14)
Kyung Mi Choi, Jeong Jin Kim, Jihye Yoo, Ku Sul Kim, Youngeun Gu, John Eom, Haengdueng Jeong, Kyungeun Kim, Ki Taek Nam, Young Soo Park, Joon-Yong Chung, Jun-Young Seo
ZUSAMMENFASSUNG
Metabolic reprogramming is an important cancer hallmark. However, the mechanisms driving metabolic phenotypes of cancer cells are unclear. Here, we showed that the interferon (IFN)-inducible protein, viperin, drives metabolic alteration in cancer cells. Viperin was observed in various types of cancer and inversely correlated with the survival rate of patients with gastric, lung, breast, renal, pancreatic, or brain cancer. By generating viperin knockdown or stably expressing cancer cells, we showed that viperin, but not a mutant lacking its iron-sulfur cluster-binding motif, increased lipogenesis and glycolysis via inhibition of fatty acid β-oxidation in cancer cells. In the tumor microenvironment, deficiency of fatty acids and oxygen as well as production of IFNs upregulated viperin expression via the PI3K/AKT/mTOR/HIF-1α and JAK/STAT pathways. Moreover, viperin was primarily expressed in cancer stem-like cells (CSCs) and functioned to promote metabolic reprogramming and enhance CSC properties, thereby facilitating tumor growth in xenograft mouse models. Collectively, our data indicate that viperin-mediated metabolic alteration drives the metabolic phenotype and progression of cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.