Merck
  • The effect of repeated periods of speed endurance training on performance, running economy, and muscle adaptations.
The effect of repeated periods of speed endurance training on performance, running economy, and muscle adaptations.
Scandinavian journal of medicine & science in sports (2017-05-26)
C Skovgaard, N W Almquist, J Bangsbo
ZUSAMMENFASSUNG
The effect of repeated intense training interventions was investigated in eight trained male runners (maximum oxygen uptake [VO2 -max]: 59.3±3.2 mL/kg/min, mean±SD) who performed 10 speed endurance training (SET; repeated 30-seconds "all-out" bouts) and 10 aerobic moderate-intensity training sessions during two 40-day periods (P1 and P2) separated by ~80 days of habitual training. Before and after both P1 and P2, subjects completed an incremental test to exhaustion to determine VO2 -max and a repeated running test at 90% vVO2 -max to exhaustion (RRT) to determine short-term endurance capacity. In addition, running economy (RE) was measured at 60% vVO2 -max (11.9±0.5 km/h) and v10-km (14.3±0.9 km/h), a 10-km track-running test was performed, and a biopsy from m. vastus lateralis was collected. 10-km performance and VO2 -max (mL/min) were the same prior to P1 and P2, whereas RE was better (P<.05) before P2 than before P1. During P1 and P2, 10-km performance (2.9% and 2.3%), VO2 -max (2.1% and 2.6%), and RE (1.9% and 1.8% at 60% vVO2 -max; 1.6% and 2.0% at v10-km) improved (P<.05) to the same extent, respectively. Performance in RRT was 20% better (P<.05) after compared to before P2, with no change in P1. No changes in muscle expression of Na+ ,K+ -ATPase α1, α2 and β1, NHE1, SERCA1 and SERCA2, actin, and CaMKII were found during neither P1 nor P2. Thus, the present study demonstrates that a second period of intense training leads to improved short-term performance and further improved RE, whereas 10-km performance and VO2 -max improve to the same extent as during the first period.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Na+/H+ Exchanger-1 Antibody, CT, clone 4E9, clone 4E9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Na+K+ ATPase α-2 Antibody, serum, Upstate®