Merck
Extraciliary roles of the ciliopathy protein JBTS17 in mitosis and neurogenesis.
Annals of neurology (2019-04-21)
Hyowon Hong, Kwangsic Joo, Sang Min Park, Jimyung Seo, Min Hwan Kim, EunBie Shin, Hae Il Cheong, Jeong Ho Lee, Joon Kim
ZUSAMMENFASSUNG
JBTS17 is a major gene mutated in ciliopathies such as Joubert syndrome and oral-facial-digital syndrome type VI. Most patients with loss of function mutations in JBTS17 exhibit cerebellar vermis hypoplasia and brainstem malformation. However, some patients with JBTS17 mutations show microcephaly and abnormal gyration. We examined potential roles of JBTS17 in neurogenesis to understand the pathological mechanism of JBTS17-related cortical abnormalities. We examined subcellular localization and cell-cycle-dependent expression of JBTS17 proteins using anti-JBTS17 antibodies and JBTS17 expression vectors. We also performed knockdown experiments to determined roles of JBTS17 in human cells, and demonstrated mitotic functions of JBTS17 using immunostaining and live imaging. We examined the involvement of JBTS17 in cortical neurogenesis using a mouse in utero electroporation technique. We found that JBTS17 localizes to the kinetochore and the level of JBTS17 is regulated by cell-cycle-dependent proteolysis. Depletion of JBTS17 disrupts chromosome alignment and spindle pole orientation, resulting in mitotic delay. JBTS17 interacts with LIS1 and influences LIS1 localization. Depletion of Jbts17 in the developing mouse cortex interferes with the mitotic progression of neural progenitors and the migration of postmitotic neurons. LIS1 is implicated in lissencephaly, but altered dosage of LIS1 has been also associated with microcephaly syndromes. Our results suggest that JBTS17 contributes to mitotic progression by interacting with LIS1, and abnormal mitosis is an underlying mechanism of the microcephaly phenotype in JBTS17-related ciliopathies. We propose that understanding extraciliary roles of ciliopathy proteins is important to elucidate pathological mechanisms underlying diverse ciliopathy phenotypes. ANN NEUROL 2019.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-C5orf42 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution