Merck
SENP3-mediated TIP60 deSUMOylation is required for DNA-PKcs activity and DNA damage repair.
MedComm (2022-04-01)
Yang Han, Xin Huang, Xiaoyu Cao, Yuchen Li, Lei Gao, Jin Jia, Gang Li, Hejiang Guo, Xiaochang Liu, Hongling Zhao, Hua Guan, Pingkun Zhou, Shanshan Gao
ZUSAMMENFASSUNG
The activation of DNA-dependent kinase (DNA-PKcs) upon DNA damage contains a cascade of reactions, covering acetylation by TIP60, binding with Ku70/80, and autophosphorylation. However, how cells regulate TIP60-mediated acetylation of DNA-PKcs and the following DNA-PKcs activation upon DNA damage remains obscure. This present study reported that TIP60 is hyper-SUMOylated in normal conditions, but upon irradiation-induced DNA damage, small ubiquitin-like modifier (SUMO)-specific protease 3 (SENP3)-mediated deSUMOylation of TIP60 promoted its interaction with DNA-PKcs to form the TIP60-DNA-PKcs complex. We show that TIP60 SUMOylation is reduced quickly in response to DNA damage and the deSUMOylation of TIP60 by SENP3 is required for DNA-PKcs acetylation and its autophosphorylation. Comet and γH2AX immunofluorescence assay showed that knockdown of SENP3 impaired DNA damage repair. Using the NHEJ report system, we found that knockdown of SENP3 affected the efficiency of NHEJ. Further exploration using clonogenic survival assay, cell viability assay and cytoflow assay suggested that leaking SENP3 increased the sensitivity of tumour cells to serval DNA damage treatment. Overall, our findings revealed a previously unidentified role of SENP3 in regulating DNA-PKcs activity and DNA damage repair.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Etoposid, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid