Merck
  • A positive allosteric modulator for the muscarinic receptor (M1 mAChR) improves pathology and cognitive deficits in female APPswe/PSEN1ΔE9 mice.
A positive allosteric modulator for the muscarinic receptor (M1 mAChR) improves pathology and cognitive deficits in female APPswe/PSEN1ΔE9 mice.
British journal of pharmacology (2021-11-26)
Khaled S Abd-Elrahman, Shaarika Sarasija, Tash-Lynn L Colson, Stephen S G Ferguson
ZUSAMMENFASSUNG
Alzheimer's disease (AD) is a neurodegenerative disease characterized by progressive cognitive decline, and women account for 60% of diagnosed cases. β-Amyloid (Aβ) oligomers are considered the principal neurotoxic species in AD brains. The M1 muscarinic ACh receptor (M1 mAChR) plays a key role in memory and learning. M1 mAChR agonists show pro-cognitive activity but cause many adverse off-target effects. A new orally bioavailable M1 mAChR positive allosteric modulator (PAM), VU0486846, is devoid of direct agonist activity or adverse effects but was not tested for disease-modifying efficacy in female AD mice. Nine-month-old female APPswe/PSEN1ΔE9 (APPswe) and wildtype mice were treated with VU0486846 in drinking water (10 mg·kg-1 ·day-1 ) for 4 or 8 weeks. Cognitive function of mice was assessed after treatment, and brains were harvested for biochemical and immunohistochemical assessment. VU0486846 improved cognitive function of APPswe mice when tested in novel object recognition and Morris water maze. This was paralleled by a significant reduction in Aβ oligomers and plaques and neuronal loss in the hippocampus. VU0486846 reduced Aβ oligomer production in APPswe mice by increasing M1 mAChR expression and shifting the processing of amyloid precursor protein from amyloidogenic cleavage to non-amyloidogenic cleavage. Specifically, VU0486846 reduced the expression of β-secretase 1 (BACE1), whereas it enhanced the expression of the α-secretase ADAM10 in APPswe hippocampus. Using M1 mAChR PAMs can be a viable disease-modifying approach that should be exploited clinically to slow AD in women.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
Protease-Inhibitor-Cocktail-Set III, EDTA-frei, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-NeuN-Antikörper (Kaninchen), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-β-Tubulin-III in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor (M1) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
VU0486846, ≥98% (HPLC)