Merck
  • An autopsy case of Alzheimer's disease presenting with primary progressive aphasia: a clinicopathological and immunohistochemical study.
An autopsy case of Alzheimer's disease presenting with primary progressive aphasia: a clinicopathological and immunohistochemical study.
Neuropathology : official journal of the Japanese Society of Neuropathology (2001-01-02)
F Li, E Iseki, M Kato, Y Adachi, M Akagi, K Kosaka
ZUSAMMENFASSUNG
This report describes an autopsied Alzheimer's disease (AD) patient with primary progressive aphasia (PPA) as an early symptom. The patient developed a progressive speech disturbance at the age of 70 years, and difficulty in comprehension became apparent 2 years later. Magnetic resonance imaging scan disclosed asymmetrical brain atrophy, predominantly on the left temporal lobe. At the age of 74 years, the patient's dementia rapidly progressed with parkinsonism and he died after a disease duration of 6 years. At autopsy, the brain showed a marked temporo-frontal lobe atrophy, predominantly on the left side. There was severe neuronal loss with gliosis and tissue rarefaction in the atrophied cerebral cortex and amygdala. Many neurofibrillary tangles with neuropil threads were found in the cerebral cortex. Numerous amyloid deposits were distributed throughout the cerebral cortex, accompanied by amyloid angiopathies. This patient was clinically diagnosed with temporal lobe-dominant Pick's disease, although the possibility of corticobasal degeneration was made. The neuropathological diagnosis was AD with asymmetrical brain atrophy and widespread amyloid angiopathies.