Merck
  • Influence of Bile Salts and Pancreatin on Dog Food during Static In Vitro Simulation to Mimic In Vivo Digestion.
Influence of Bile Salts and Pancreatin on Dog Food during Static In Vitro Simulation to Mimic In Vivo Digestion.
Animals : an open access journal from MDPI (2022-10-28)
Kangmin Seo, Hyun-Woo Cho, Jung-Hwan Jeon, Chan Ho Kim, Sejin Lim, Sohee Jeong, Kihyun Kim, Ju Lan Chun
ZUSAMMENFASSUNG
The addition of pancreatin and bile salts in different concentrations during in vitro digestion causes changes in the digestibility of crude protein (CP), fat, and dry matter (DM). The effects of bile salts and pancreatin on the digestibility of ether extract (EE), CP, and DM in developing a static in vitro digestion model for dogs were assessed using different concentrations of pancreatin (0, 1, 2.5, 5, and 10 g/L digestive solution) and bile salts (0, 2.5, 6.25, 12.5, and 25 g/L digestive solution). The data were analyzed using one-way analysis of variance. Digestibility of EE increased with the addition of bile salts (p < 0.05), whereas that of CP decreased with ≤0.25 g (1.0 g/L digestive solution) pancreatin. The digestibility of DM decreased significantly in all groups supplemented with ≥3.125 g (12.5 g/L digestive solution) bile salts and 0.25-2.5 g (1-10 g/L digestive solution) pancreatin and was the lowest with 6.25 g (25 g/L digestive solution) of bile salts (p < 0.05). These findings could facilitate the development of effective static in vitro digestion models for dogs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pepsin aus Schweinemagenschleimhaut, powder, ≥250 units/mg solid
Sigma-Aldrich
Pancreatin aus Schweinepankreas, 8 × USP specifications