Merck
  • Synthesis and biological evaluation of kojic acid derivatives containing 1,2,4-triazole as potent tyrosinase inhibitors.
Synthesis and biological evaluation of kojic acid derivatives containing 1,2,4-triazole as potent tyrosinase inhibitors.
Chemical biology & drug design (2015-04-29)
Wenlin Xie, Jingai Zhang, Xiaojing Ma, Wenqian Yang, Ying Zhou, Xufu Tang, Yan Zou, Hui Li, Jingjing He, Shimin Xie, Yunhui Zhao, Fengping Liu
ZUSAMMENFASSUNG
A series of 5-substituted-3-[5-hydroxy-4-pyrone-2-yl-methymercapto]-4-amino-1,2,4-triazole derivatives were synthesized by nucleophilic substitution reaction of 5-hydroxy-2-chloromethyl -4H-pyran-4-one with 5-substituted-3-mercapto-4-amino-1,2,4-triazole, and their inhibitory effects on mushroom tyrosinase were evaluated. The results indicated that most of the synthesized compounds exhibited significant inhibitory activity. Specifically, 5-(4-chlorophenyl)-3-[5-hydroxy-4-pyrone-2-yl-methymercapto]-4-amino-1,2,4-triazole (6j) exhibited the most potent tyrosinase inhibitory activity with IC50 value of 4.50 ± 0.34 μm. The kinetic studies of the compound (6j) demonstrated that the inhibitory effects of the compound on the tyrosinase were belonging to competitive inhibitors. Meanwhile, the structure-activity relationship was also discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyrosinase aus Ständerpilz, lyophilized powder, ≥1000 unit/mg solid