- Derivation and characterization of neuronal precursors and dopaminergic neurons from human embryonic stem cells in vitro.
Derivation and characterization of neuronal precursors and dopaminergic neurons from human embryonic stem cells in vitro.
Methods in molecular biology (Clifton, N.J.) (2006-08-03)
Melissa Carpenter, Mahendra S Rao, William Freed, Xianmin Zeng
PMID16881516
ZUSAMMENFASSUNG
Dopaminergic neurons of human origin have many potential research applications such as in vitro studies on biochemical pathways related to neuronal disorders, and potentially direct cell replacement for therapeutic use. Dopaminergic neurons with apparently normal properties can be produced from embryonic stem cells from mice and sub-human primates by the simple procedure of coculturing with the PA6 stromal cell line. Recently, we have demonstrated that this coculture system can induce dopaminergic differentiation in human embryonic stem cells, and the human embryonic stem cell-derived dopaminergic cells exhibit biochemical and functional properties of mature dopaminergic neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoklonaler Anti-β-Tubulin-III-Antikörper in Maus hergestellte Antikörper, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Neural Cell Adhesion Molecule antibody produced in mouse, clone NCAM-0B11, ascites fluid