Merck
Detection of Long Non-coding RNA Expression by Non-radioactive Northern Blots.
Methods in molecular biology (Clifton, N.J.) (2021-08-22)
Yueying Wang, Mu Xu, Jiao Yuan, Zhongyi Hu, Youyou Zhang, Lin Zhang, Xiaowen Hu
ZUSAMMENFASSUNG
With the advances in sequencing technology and transcriptome analysis, it is estimated that up to 75% of the human genome is transcribed into RNAs. This finding prompted intensive investigations on the biological functions of non-coding RNAs and led to very exciting discoveries of microRNAs as important players in disease pathogenesis and therapeutic applications. Research on long non-coding RNAs (lncRNAs) is in its infancy, yet a broad spectrum of biological regulations has been attributed to lncRNAs. As a novel class of RNA transcripts, the expression level and splicing variants of lncRNAs are various. Northern blot analysis can help us learn about the identity, size, and abundance of lncRNAs. Here we describe how to use northern blot to determine lncRNA abundance and identify different splicing variants of a given lncRNA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-Digoxigenin-AP, Fab-Fragmente, from sheep