Merck
  • Synthetic identification as a hexapeptide of alpha substance-IB inducing sexual agglutination in Saccharomyces cerevisiae.
Synthetic identification as a hexapeptide of alpha substance-IB inducing sexual agglutination in Saccharomyces cerevisiae.
Experientia (1977-07-15)
H Aoyagi, N Izumiya, A Sakurai, K Sakata, S Tamura
PMID330197
ZUSAMMENFASSUNG
For the identification of a peptidyl principle inducing sexual agglutination in the yeast, 2 supposed hexapeptides (1a, b) were synthesized by the conventional method. The 1a (H-Arg-Gly-Pro-Phe-Pro-Ile-OH) revealed complete identity with the natural peptide in TLC, MS and biological property on agglutination. The 1b showed the sexual agglutinability in the same degree as 1a, though distinct differences were observed in the chemical data. Both 1a and 1b had a strong bitter taste.