Merck
  • Prolidase enzyme is required for extracellular matrix integrity and impacts on postnatal cerebellar cortex development.
Prolidase enzyme is required for extracellular matrix integrity and impacts on postnatal cerebellar cortex development.
The Journal of comparative neurology (2019-06-28)
Violetta Insolia, Erica C Priori, Caterina Gasperini, Federica Coppa, Marco Cocchia, Erika Iervasi, Beatrice Ferrari, Roberta Besio, Silvia Maruelli, Graziella Bernocchi, Antonella Forlino, Maria G Bottone
ZUSAMMENFASSUNG
The extracellular matrix is essential for brain development, lamination, and synaptogenesis. In particular, the basement membrane below the pial meninx (pBM) is required for correct cortical development. The last step in the catabolism of the most abundant protein in pBM, collagen Type IV, requires prolidase, an exopeptidase cleaving the imidodipeptides containing pro or hyp at the C-terminal end. Mutations impairing prolidase activity lead in humans to the rare disease prolidase deficiency characterized by severe skin ulcers and mental impairment. Thus, the dark-like (dal) mouse, in which the prolidase is knocked-out, was used to investigate whether the deficiency of prolidase affects the neuronal maturation during development of a brain cortex area. Focusing on the cerebellar cortex, thinner collagen fibers and disorganized pBM were found. Aberrant cortical granule cell proliferation and migration occurred, associated to defects in brain lamination, and in particular in maturation of Purkinje neurons and formation of synaptic contacts. This study deeply elucidates a link between prolidase activity and neuronal maturation shedding new light on the molecular basis of functional aspects in the prolidase deficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoklonaler Anti-Calbindin-D-28K-Antikörper in Maus hergestellte Antikörper, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-VGluT2-Antikörper, serum, from guinea pig
Sigma-Aldrich
Anti-GABA A-Rezeptor β2/3-Antikörper, clone 62-3G1, Upstate®, from mouse