Merck
  • Suppression of amyloid-β secretion from neurons by cis-9, trans-11-octadecadienoic acid, an isomer of conjugated linoleic acid.
Suppression of amyloid-β secretion from neurons by cis-9, trans-11-octadecadienoic acid, an isomer of conjugated linoleic acid.
Journal of neurochemistry (2021-08-12)
Saori Hata, Kuniyuki Kano, Kazunori Kikuchi, Shoichi Kinoshita, Yuriko Sobu, Haruka Saito, Takashi Saito, Takaomi C Saido, Yoshitake Sano, Hidenori Taru, Junken Aoki, Hiroto Komano, Taisuke Tomita, Shunji Natori, Toshiharu Suzuki
ZUSAMMENFASSUNG
Two common conjugated linoleic acids (LAs), cis-9, trans-11 CLA (c9,t11 CLA) and trans-10, cis-12 CLA (t10,c12 CLA), exert various biological activities. However, the effect of CLA on the generation of neurotoxic amyloid-β (Aβ) protein remains unclear. We found that c9,t11 CLA significantly suppressed the generation of Aβ in mouse neurons. CLA treatment did not affect the level of β-site APP-cleaving enzyme 1 (BACE1), a component of active γ-secretase complex presenilin 1 amino-terminal fragment, or Aβ protein precursor (APP) in cultured neurons. BACE1 and γ-secretase activities were not directly affected by c9,t11 CLA. Localization of BACE1 and APP in early endosomes increased in neurons treated with c9,t11 CLA; concomitantly, the localization of both proteins was reduced in late endosomes, the predominant site of APP cleavage by BACE1. The level of CLA-containing phosphatidylcholine (CLA-PC) increased dramatically in neurons incubated with CLA. Incorporation of phospholipids containing c9,t11 CLA, but not t10,c12 CLA, into the membrane may affect the localization of some membrane-associated proteins in intracellular membrane compartments. Thus, in neurons treated with c9,t11 CLA, reduced colocalization of APP with BACE1 in late endosomes may decrease APP cleavage by BACE1 and subsequent Aβ generation. Our findings suggest that the accumulation of c9,t11 CLA-PC/LPC in neuronal membranes suppresses the production of neurotoxic Aβ in neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
5-Fluor-2′-Desoxyuridin, thymidylate synthase inhibitor