Merck
  • Synthesis and biological evaluation of novel N-aryl-ω-(benzoazol-2-yl)-sulfanylalkanamides as dual inhibitors of α-glucosidase and protein tyrosine phosphatase 1B.
Synthesis and biological evaluation of novel N-aryl-ω-(benzoazol-2-yl)-sulfanylalkanamides as dual inhibitors of α-glucosidase and protein tyrosine phosphatase 1B.
Chemical biology & drug design (2018-05-11)
Mei-Yan Wang, Xian-Chao Cheng, Xiu-Bo Chen, Yu Li, Lan-Lan Zang, Yu-Qing Duan, Ming-Zhu Chen, Peng Yu, Hua Sun, Run-Ling Wang
ZUSAMMENFASSUNG
α-Glucosidase is known to catalyze the digestion of carbohydrates and release free glucose into the digestive tract. Protein tyrosine phosphatase 1B (PTP1B) is engaged in the dephosphorylation of the insulin receptor and regulation of insulin sensitivity. Therefore, dual antagonists by targeting both α-glucosidase and PTP1B may be potential candidates for type 2 diabetes therapy. In this work, three series of novel N-aryl-ω-(benzoazol-2-yl)-sulfanylalkanamides were synthesized and assayed for their α-glucosidase and PTP1B inhibitory activities, respectively. Compound 3l, exhibiting the most effective α-glucosidase inhibitory activity (IC50  = 10.96 μm (3l), IC50  = 51.32 μm (Acarbose), IC50  = 18.22 μm (Ursolic acid)) and potent PTP1B inhibitory activity (IC50  = 13.46 μm (3l), IC50  = 14.50 μm (Ursolic acid)), was identified as a novel dual inhibitor of α-glucosidase and PTP1B. Furthermore, 3l is a highly selective PTP1B inhibitor because no inhibition was showed by 3l at 100 μm against PTP-MEG2, TCPTP, SHP2, or SHP1. Subsequent kinetic analysis revealed 3l inhibited α-glucosidase in a reversible and mixed manner. Molecular docking study indicated that hydrogen bonds, van der Waals, charge interactions and Pi-cation interactions all contributed to affinity between 3l and α-glucosidase/PTP1B.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Glucosidase aus Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)