Merck
  • 3-Aryl Coumarin Derivatives Bearing Aminoalkoxy Moiety as Multi-Target-Directed Ligands against Alzheimer's Disease.
3-Aryl Coumarin Derivatives Bearing Aminoalkoxy Moiety as Multi-Target-Directed Ligands against Alzheimer's Disease.
Chemistry & biodiversity (2019-04-09)
Helia Abdshahzadeh, Mostafa Golshani, Hamid Nadri, Iraj Saberi Kia, Zahra Abdolahi, Hamid Forootanfar, Alieh Ameri, Tuba Tüylü Küçükkılınç, Beyza Ayazgok, Leili Jalili-Baleh, Seyed Esmaeil Sadat Ebrahimi, Setareh Moghimi, Ismaeil Haririan, Mehdi Khoobi, Alireza Foroumadi
ZUSAMMENFASSUNG
Two series of novel coumarin derivatives, substituted at 3 and 7 positions with aminoalkoxy groups, are synthesized, characterized, and screened. The effect of amine substituents and the length of cross-linker are investigated in acetyl- and butyrylcholinesterase (AChE and BuChE) inhibition. Target compounds show moderate to potent inhibitory activities against AChE and BuChE. 3-(3,4-Dichlorophenyl)-7-[4-(diethylamino)butoxy]-2H-chromen-2-one (4y) is identified as the most potent compound against AChE (IC50 =0.27 μm). Kinetic and molecular modeling studies affirmed that compound 4y works in a mixed-type way and interacts simultaneously with the catalytic active site (CAS) and peripheral anionic site (PAS) of AChE. In addition, compound 4y blocks β-amyloid (Aβ) self-aggregation with a ratio of 44.11 % at 100 μm and significantly protects PC12 cells from H2 O2 -damage in a dose-dependent manner.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amyloid-β-Proteinfragment 1-42
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (BCA)