Merck
Cell behavior on surface modified polydimethylsiloxane (PDMS).
Macromolecular bioscience (2014-03-07)
Morgan M Stanton, Johanna M Rankenberg, Byung-Wook Park, W Grant McGimpsey, Christopher Malcuit, Christopher R Lambert
ZUSAMMENFASSUNG
Designing complex tissue culture systems requires cell alignment and directed extracellular matrix (ECM) and gene expression. Here, a micro-rough, polydimethylsiloxane (PDMS) surface, that also integrates a micro-pattern of 50 µm wide lines of fibronectin (FN) separated by 60 µm wide lines of bovine serum albumin (BSA), is developed. Human fibroblasts cultured on the rough, patterned substrate have aligned growth and a significant change in morphology when compared to cells on a flat, patterned surface. The rough PDMS topography significantly decreases cell area and induces the upregulation of several ECM related genes by two-fold when compared to cells cultured on flat PDMS. This study describes a simple surface engineering procedure for creating surface architecture for scaffolds to design and control the cell-surface interface.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronectin-Humanplasma, lyophilized powder, BioReagent, suitable for cell culture