Merck
Macromolecular Coating Enables Tunable Selectivity in a Porous PDMS Matrix.
Macromolecular bioscience (2017-12-23)
Benjamin Winkeljann, Benjamin T Käsdorf, Job Boekhoven, Oliver Lieleg
ZUSAMMENFASSUNG
Whether for laboratory use or clinical practice, many fields in Life Sciences require selective filtering. However, most existing filter systems lack the ability to easily tune their filtration behavior. Two key elements for efficient filtering are a high surface-to-volume ratio and the presence of suitable chemical groups which establish selectivity. In this study, an artificial PDMS-based capillary system with highly tunable selectivity properties is presented. The high surface-to-volume ratio of this filter system is generated by first embedding sugar fibers into a synthetic polymer matrix and then dissolving these fibers from the cured polymer. To functionalize this filter, the inner surface of the capillaries is coated with purified or synthetic macromolecules. Depending on the type of macromolecule used for filter functionalization, selective sieving is observed based on steric hindrance, electrostatic binding, electrostatic repulsion, or specific binding interactions. Furthermore, it is demonstrated that enzymes can be immobilized in the capillary system which allows for performing multiple cycles of enzymatic reactions with the same batch of enzymes and without the need to separate the enzymes from their reaction products. In addition to lab-scale filtration and enzyme immobilization applications demonstrated here, the functionalized porous PDMS matrix may also be used to test binding interactions between different molecules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mucin aus Schweinemagen, Type II