Merck
  • Increased expression of extracellular matrix metalloproteinase inducer (EMMPRIN) and MMP10, MMP23 in inflammatory bowel disease: Cross-sectional study.
Increased expression of extracellular matrix metalloproteinase inducer (EMMPRIN) and MMP10, MMP23 in inflammatory bowel disease: Cross-sectional study.
Scandinavian journal of immunology (2020-08-28)
Gabriela Fonseca-Camarillo, Janette Furuzawa-Carballeda, Braulio Martínez-Benitez, Rafael Barreto-Zuñiga, Jesús K Yamamoto-Furusho
ZUSAMMENFASSUNG
It has been reported that EMMPRIN is involved in the regulation of immune response and the induction of MMPs production by fibroblasts. The aim of this study was to describe the intestinal gene expression and protein production of EMMPRIN, MMP23 and MMP10 in patients with ulcerative colitis (UC) and Crohn's disease (CD) and compared them with a control group. Gene expression of EMMPRIN, MMP10 and MMP23B was measured by RT-PCR. In order to determine EMMPRIN and MMP protein expression, colonic tissues were immunostained. The results of the study showed EMMPRIN gene expression was upregulated in rectal mucosa from active (a)UC versus aCD patients (P = .045), remission (r)CD group (P = .0009) and controls (P < .0001). We detected differences between rUC and aCD (P = .004), rCD (P < .0001) or control group (P < .0001). EMMPRIN showed a higher expression in mucosa (intraepithelial lymphocytes), submucosa and adventitia (endothelial cells) from aCD patients. MMP23 levels were increased in aUC and aCD compared to rUC and rCD and the control group (P = .0001). EMMPRIN+/MMP23+─expressing cells were localized mainly in mucosa, muscular and adventitia from active UC patients. MMP10 gene expression was increased in aUC versus CD patients and the control group (P = .0001). MMP10 gene expression is associated with inflammation in UC patients (P = .0001, r2  = .585). EMMPRIN+/MMP10+─producing cells were found mainly in all intestinal layers and perivascular inflammatory infiltrates from aUC patients. In conclusion, EMMPRIN, MMP23 and MMP10 were upregulated in patients with active UC versus remission UC , CD and control groups suggesting that, they are involved in the inflammatory process.