- Mouse Embryonic Stem Cell Culture in Serum-Containing or 2i Conditions.
Mouse Embryonic Stem Cell Culture in Serum-Containing or 2i Conditions.
Methods in molecular biology (Clifton, N.J.) (2021-10-19)
Emre Balbasi, Gozde Guven, Nihal Terzi Cizmecioglu
PMID34661879
ZUSAMMENFASSUNG
With their unique capabilities of self-renewal and differentiation into three germ layers, mouse embryonic stem cells (mESCs) are widely used as an in vitro cellular model for early mammalian developmental studies. mESCs are traditionally cultured in high-serum and LIF-containing medium on a growth-deficient mouse embryonic fibroblast layer. A more recent culturing system with two inhibitors (for GSK3β (CHIR99021) and MEK1/2 (PD0325901)) and LIF enables the derivation of mESC lines from various mouse strains. Here we describe methods for the mESC growth and maintenance in each medium composition as well as their adaptation to either condition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ESGRO® Rekombinantes Maus-LIF-Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.