Merck
Quantification of protein carbonylation.
Methods in molecular biology (Clifton, N.J.) (2013-01-09)
Nancy B Wehr, Rodney L Levine
ZUSAMMENFASSUNG
Protein carbonylation is the most commonly used measure of oxidative modification of proteins. It is most often measured spectrophotometrically or immunochemically by derivatizing proteins with the classical carbonyl reagent 2,4 dinitrophenylhydrazine (DNPH). We present protocols for the derivatization and quantification of protein carbonylation with these two methods, including a newly described dot blot with greatly increased sensitivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Dinitrophenyl-Antikörper, Maus monoklonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution