Merck
Methods for Tumor Targeting with Salmonella typhimurium A1-R.
Methods in molecular biology (Clifton, N.J.) (2016-02-06)
Robert M Hoffman, Ming Zhao
ZUSAMMENFASSUNG
Salmonella typhimurium A1-R (S. typhimurium A1-R) has shown great preclinical promise as a broad-based anti-cancer therapeutic (please see Chapter 1 ). The present chapter describes materials and methods for the preclinical study of S. typhimurium A1-R in clinically-relevant mouse models. Establishment of orthotopic metastatic mouse models of the major cancer types is described, as well as other useful models, for efficacy studies of S. typhimurium A1-R or other tumor-targeting bacteria, as well. Imaging methods are described to visualize GFP-labeled S. typhimurium A1-R, as well as GFP- and/or RFP-labeled cancer cells in vitro and in vivo, which S. typhimurium A1-R targets. The mouse models include metastasis to major organs that are life-threatening to cancer patients including the liver, lung, bone, and brain and how to target these metastases with S. typhimurium A1-R. Various routes of administration of S. typhimurium A1-R are described with the advantages and disadvantages of each. Basic experiments to determine toxic effects of S. typhimurium A1-R are also described. Also described are methodologies for combining S. typhimurium A1-R and chemotherapy. The testing of S. typhimurium A1-R on patient tumors in patient-derived orthotopic xenograft (PDOX) mouse models is also described. The major methodologies described in this chapter should be translatable for clinical studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-PECAM-1-Antikörper, Klon 390, clone 390, Chemicon®, from rat