Merck
  • In Vitro Models for Studying the Interaction of Neisseria meningitidis with Human Brain Endothelial Cells.
In Vitro Models for Studying the Interaction of Neisseria meningitidis with Human Brain Endothelial Cells.
Methods in molecular biology (Clifton, N.J.) (2019-03-17)
Brandon J Kim, Alexandra Schubert-Unkmeir
ZUSAMMENFASSUNG
Bacterial meningitis is a serious, life-threatening infection of the central nervous system (CNS). To cause meningitis, bacteria must interact with and penetrate the meningeal blood-cerebrospinal fluid barrier (mB/CSFB), which comprises highly specialized brain endothelial cells. Neisseria meningitidis (meningococcus) is a leading cause of bacterial meningitis, and examination meningococcus' interaction with the BBB is critical for understanding disease progression. To examine specific interactions, in vitro mB/CSFB models have been developed and employed and are of great importance because in vivo models have been difficult to produce considering Neisseria meningitidis is exclusively a human pathogen. Most in vitro blood-brain barrier and mB/CSF models use primary and immortalized brain endothelial cells, and these models have been used to examine bacterial-mB/CSFB interactions by a variety of pathogens. This chapter describes the use of past and current in vitro brain endothelial cells to model Neisseria meningitidis interaction with the mB/CSFB, and inform on the standard operating procedure for their use.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Blut-Hirn-Schranken-hCMEC/D3-Zelllinie, The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.
Sigma-Aldrich
Anti-FAK-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution