Merck
Interaction of Alt a 1 with SLC22A17 in the airway mucosa.
Allergy (2019-05-17)
María Garrido-Arandia, Jaime Tome-Amat, Diego Pazos-Castro, Vanesa Esteban, Maria M Escribese, Guadalupe Hernández-Ramírez, Alma Yuste-Montalvo, Domingo Barber, Luis F Pacios, Araceli Díaz-Perales
ZUSAMMENFASSUNG
Despite all the efforts made up to now, the reasons that facilitate a protein becoming an allergen have not been elucidated yet. Alt a 1 protein is the major fungal allergen responsible for chronic asthma, but little is known about its immunological activity. Our main purpose was to investigate the ligand-dependent interactions of Alt a 1 in the human airway epithelium. Alt a 1 with and without its ligand (holo- and apo- forms) was incubated with the pulmonary epithelial monolayer model, Calu-3 cells. Allergen transport and cytokine production were measured. Pull-down and immunofluorescence assays were employed to identify the receptor of Alt a 1 using the epithelial cell model and mouse tissues. Receptor-allergen-ligand interactions were analyzed by computational modeling. The holo-form could activate human monocytes, PBMCs, and polarized airway epithelial (Calu-3) cell lines. The allergen was also transported through the monolayer, without any alteration of the epithelial integrity (TEER). Alt a 1 also induced the production of proinflammatory IL8 and specific epithelial cytokines (IL33 and IL25) by Calu-3 cells. The interaction between epithelial cells and holo-Alt a 1 was found to be mediated by the SLC22A17 receptor, and its recognition of Alt a 1 was explained in structural terms. Our findings identified the Alt a 1 ligand as a central player in the interaction of the allergen with airway mucosa, shedding light into its potential role in the immunological response, while unveiling its potential as a new target for therapy intervention.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid