Merck
  • Sirtuin 3 is required for the protective effect of Resveratrol on Manganese-induced disruption of mitochondrial biogenesis in primary cultured neurons.
Sirtuin 3 is required for the protective effect of Resveratrol on Manganese-induced disruption of mitochondrial biogenesis in primary cultured neurons.
Journal of neurochemistry (2020-05-20)
Qian Sun, Run-Run Kang, Kai-Ge Chen, Kuan Liu, Zhuo Ma, Chang Liu, Yu Deng, Wei Liu, Bin Xu
ZUSAMMENFASSUNG
Chronic manganese (Mn) exposure can disturb mitochondrial homeostasis leading to mitochondrial dysfunction, which is involved in Mn-induced neurodegenerative diseases. Resveratrol (RSV), as a promoter of mitochondrial biogenesis, plays a significant role against mitochondrial dysfunction. However, whether RSV can relieve Mn-induced neuronal injury and mitochondrial dysfunction remains unknown. Sirtuin 3 (SIRT3), a main mitochondrial sirtuin, is an important regulator of mitochondria to maintain mitochondrial homeostasis. Therefore, this study investigated whether SIRT3 was required for RSV alleviating Mn-induced mitochondrial dysfunction in primary cultured neurons from C57BL/6 mice. Here, we showed that Mn (100 and 200 μM) exposure for 24 hr caused significant neuronal damage and mitochondrial dysfunction through increasing mitochondrial ROS, reducing mitochondrial membrane potential and adenosine triphosphate level, and leading to mitochondrial network fragmentation, which could be ameliorated by RSV pretreatment in primary cultured neurons. Additionally, our results also indicated that RSV could activate the SIRT1/PGC-1α signaling pathway and alleviate Mn-induced disruption of mitochondrial biogenesis by increasing SIRT1 expression and activity, enhancing deacetylation of PGC-1α. Furthermore, SIRT3 over-expression increased deacetylation of mitochondrial transcription factor A and mitochondrial DNA (mtDNA) copy number. Oppositely, silencing SIRT3 increased acetylation of mitochondrial transcription factor A and decreased mtDNA copy number. Our results showed SIRT3 was required for the protective effect of RSV in mitochondrial biogenesis. In conclusion, our findings demonstrated that RSV could ameliorate Mn-induced neuronal injury and mitochondrial dysfunction in primary cultured neurons through activating the SIRT1/ PGC-1α signaling pathway, and that SIRT3 is required for promoting mitochondrial biogenesis and attenuating Mn-induced mitochondrial dysfunction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid, ≥90% (HPLC), crystalline