Merck
  • 6-Shogaol (enexasogoal) treatment improves experimental knee osteoarthritis exerting a pleiotropic effect over immune innate signalling responses in chondrocytes.
6-Shogaol (enexasogoal) treatment improves experimental knee osteoarthritis exerting a pleiotropic effect over immune innate signalling responses in chondrocytes.
British journal of pharmacology (2022-06-28)
Paula Gratal, Aránzazu Mediero, Ana Lamuedra, Alejandra Matamoros-Recio, Carmen Herencia, Gabriel Herrero-Beaumont, Sonsoles Martín-Santamaría, Raquel Largo
ZUSAMMENFASSUNG
The pathogenesis of osteoarthritis implicates a low-grade inflammation associated to the innate immune system activation. Toll like receptor (TLR) stimulation triggers the release of inflammatory mediators, which aggravate osteoarthritis. We studied the preventive effect of 6-shogaol, a potential TLR4 inhibitor, on the treatment of experimental knee osteoarthritis. Osteoarthritis was induced in C57BL6 mice by surgical section of the medial meniscotibial ligament, which received 6-shogaol for eight weeks. Cartilage damage, inflammatory mediator presence and disease markers were assessed in joint tissues by immunohistochemistry. Computational modelling was used to predict binding modes of 6-shogaol into the TLR4/MD2 receptor and its permeability across cellular membranes. Employing LPS-stimulated chondrocytes and MAPK assay, we elucidated 6-shogaol action mechanisms. 6-Shogaol treatment prevented articular cartilage lesions, synovitis and the presence of pro-inflammatory mediators, and disease markers in osteoarthritis animals. Molecular modelling studies predicted 6-shogaol interaction with the TLR4/MD-2 heterodimer in an antagonist conformation through its binding into the MD-2 pocket. In cell culture, we confirmed that 6-shogaol reduced LPS-induced TLR4 inflammatory signalling pathways. Besides, MAPK assay demonstrated that 6-shogaol directly inhibits the ERK1/2 phosphorylation activity. 6-Shogaol evoked a preventive action on cartilage and synovial inflammation in osteoarthritis mice. 6-shogaol effect may take place not only by hindering the interaction between TLR4 ligands and the TLR4/MD-2 complex in chondrocytes, but also through inhibition of ERK phosphorylation, implying a pleiotropic effect on different mediators activated during osteoarthritis, which proposes it as an attractive drug for osteoarthritis treatments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2