Merck
Nuclear stabilization of p53 requires a functional nucleolar surveillance pathway.
Cell reports (2022-11-03)
Katherine M Hannan, Priscilla Soo, Mei S Wong, Justine K Lee, Nadine Hein, Perlita Poh, Kira D Wysoke, Tobias D Williams, Christian Montellese, Lorey K Smith, Sheren J Al-Obaidi, Lorena Núñez-Villacís, Megan Pavy, Jin-Shu He, Kate M Parsons, Karagh E Loring, Tess Morrison, Jeannine Diesch, Gaetan Burgio, Rita Ferreira, Zhi-Ping Feng, Cathryn M Gould, Piyush B Madhamshettiwar, Johan Flygare, Thomas J Gonda, Kaylene J Simpson, Ulrike Kutay, Richard B Pearson, Christoph Engel, Nicholas J Watkins, Ross D Hannan, Amee J George
ZUSAMMENFASSUNG
The nucleolar surveillance pathway monitors nucleolar integrity and responds to nucleolar stress by mediating binding of ribosomal proteins to MDM2, resulting in p53 accumulation. Inappropriate pathway activation is implicated in the pathogenesis of ribosomopathies, while drugs selectively activating the pathway are in trials for cancer. Despite this, the molecular mechanism(s) regulating this process are poorly understood. Using genome-wide loss-of-function screens, we demonstrate the ribosome biogenesis axis as the most potent class of genes whose disruption stabilizes p53. Mechanistically, we identify genes critical for regulation of this pathway, including HEATR3. By selectively disabling the nucleolar surveillance pathway, we demonstrate that it is essential for the ability of all nuclear-acting stresses, including DNA damage, to induce p53 accumulation. Our data support a paradigm whereby the nucleolar surveillance pathway is the central integrator of stresses that regulate nuclear p53 abundance, ensuring that ribosome biogenesis is hardwired to cellular proliferative capacity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Fetales Kälberserum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture