Merck
Detection of cytoplasmic and nuclear functions of mTOR by fractionation.
Methods in molecular biology (Clifton, N.J.) (2011-11-30)
Margit Rosner, Markus Hengstschläger
ZUSAMMENFASSUNG
Subcellular localization constitutes the environment in which proteins act. It tightly controls access to and availability of different types of molecular interacting partners and is therefore a major determinant of protein function and regulation. Originally thought to be a mere cytoplasmic kinase the mammalian target of rapamycin (mTOR) has recently been localized to various intracellular compartments including the nucleus and specific components of the endomembrane system such as lysosomes. The identification of essential binding partners and the structural and functional partitioning of mTOR into two distinct multiprotein complexes warrant the detailed investigation of the subcellular localization of mTOR as part of mTORC1 and mTORC2. Upon establishment of experimental conditions allowing cytoplasmic/nuclear fractionation at high purity and maximum mTOR complex recovery we have previously shown that the mTOR/raptor complex (mTORC1) is predominantly cytoplasmic whereas the mTOR/rictor complex (mTORC2) is abundant in both compartments. Moreover, the mTORC2 complex components rictor and sin1 are dephosphorylated and dynamically distributed between the cytoplasm and the nucleus upon long-term treatment with the mTOR-inhibitor rapamycin. These findings further demonstrate that the here presented and detailly described fractionation procedure is a valuable tool to study protein localization and cytoplasmic/nuclear protein shuttling in the context of expanding mTOR signalling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®