Merck
  • Allergen specificity of IgG(4)-expressing B cells in patients with grass pollen allergy undergoing immunotherapy.
Allergen specificity of IgG(4)-expressing B cells in patients with grass pollen allergy undergoing immunotherapy.
The Journal of allergy and clinical immunology (2012-05-16)
Louisa K James, Holly Bowen, Rosaleen A Calvert, Tihomir S Dodev, Mohamed H Shamji, Andrew J Beavil, James M McDonnell, Stephen R Durham, Hannah J Gould
ZUSAMMENFASSUNG
Serum IgG(4) responses to allergen immunotherapy are well documented as blocking allergen binding to receptor-bound IgE on antigen-presenting cells and effector cells, but the molecular characteristics of treatment-induced IgG(4), particularly in relation to expressed antibody, are poorly defined. We aimed to clone and express recombinant IgG(4) from patients receiving grass pollen immunotherapy using single B cells to obtain matched heavy- and light-chain pairs. IgG(4)(+) B cells were enriched from blood samples taken from 5 patients receiving grass pollen immunotherapy. Matched heavy- and light-chain variable-region sequences were amplified from single IgG(4)(+) B cells. Variable regions were cloned and expressed as recombinant IgG(4). Binding analysis of grass pollen-specific IgG(4) was performed by using surface plasmon resonance. Functional assays were used to determine IgE blocking activity. In a separate experiment grass pollen-specific antibodies were depleted from serum samples to determine the proportion of grass pollen-specific IgG(4) within total IgG(4). Depletion of grass pollen-specific antibodies from serum led to a modest reduction in total IgG(4) levels. Matched heavy- and light-chain sequences were cloned from single IgG(4)(+) B cells and expressed as recombinant IgG(4). We identified an IgG(4) that binds with extremely high affinity to the grass pollen allergen Phl p 7. Furthermore, we found that a single specific mAb can block IgE-mediated facilitated allergen presentation, as well as IgE-mediated basophil activation. Although increases in IgG(4) levels cannot be wholly accounted for within the allergen-specific fraction, allergen immunotherapy might result in the production of high-affinity allergen-specific blocking IgG(4).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Human IgG1−Biotin antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonaler ANTI-Human-IgG3, clone HP-6050, ascites fluid
Sigma-Aldrich
Monoklonaler ANTI-Human-IgG2-Antikörper, clone HP-6014, ascites fluid