Merck
  • Autophagic degradation of claudin-5 mediated by its binding to a Clostridium perfringens enterotoxin fragment modulates endothelial barrier permeability.
Autophagic degradation of claudin-5 mediated by its binding to a Clostridium perfringens enterotoxin fragment modulates endothelial barrier permeability.
FEBS letters (2022-02-15)
Panpan Lin, Rongbang Tan, Ping Yu, Yanyu Li, Yuqian Mo, Wen Li, Jingjing Zhang
ZUSAMMENFASSUNG
The blood-brain barrier (BBB) protects the central nervous system (CNS) from harmful elements, while it also restricts efficient drug delivery into the CNS. Previously, we generated a mutated fragment of Clostridium perfringens enterotoxin (cCPEYWSH ) which specifically binds to the endothelial tight junction protein claudin-5. Here, we explore the mechanisms regulating the dynamics of membranous claudin-5 and BBB permeability. Following cCPEYWSH binding to claudin-5, caveolin-1 mediates the redistribution of claudin-5 to the cytosol. This abnormal cytosolic aggregation triggers the autophagic degradation of claudin-5, leading to an increase in BBB permeability. Enhancement or inhibition of autophagy accelerates or inhibits the degradation of cytosolic claudin-5, respectively. Our findings may pave the way for improving BBB permeability for drug delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroquin -diphosphat (Salz), powder or crystals, 98.5-101.0% (EP), meets EP testing specifications