Merck
Elastinolytic and proteolytic enzymes.
Methods in molecular biology (Clifton, N.J.) (2014-05-14)
Efrat Kessler, Mary Safrin
ZUSAMMENFASSUNG
Pseudomonas aeruginosa secretes into its environment at least seven extracellular proteases: pseudolysin (LasB protease; elastase), aeruginolysin (alkaline proteinase), staphylolysin (staphylolytic endopeptidase; LasA protease), lysyl endopeptidase (protease IV; PrpL), PASP (P. aeruginosa small protease), LepA (Large ExoProtease A), and an aminopeptidase. Their action on host proteins, both individually and synergistically, plays important roles in pathogenesis of P. aeruginosa infections. Methods to measure/detect their activities are fundamental for understanding their physiological functions, roles in pathogenesis, mechanisms of action, regulation, and secretion. Most assays for determination/detection of proteolytic activity employ modified/non-modified casein or gelatin as substrates. In the quantitative assay, fragments generated from azocasein are separated from undigested substrate by trichloroacetic acid precipitation and their absorbance is measured. In non-quantitative assays, proteolytic activity is detected as clearing zones around bacterial growth or samples of culture supernatants on casein containing solid media formed due to local casein degradation. In zymography, individual proteases are detected as clear bands in gelatin/casein containing gels after SDS-PAGE separation, renaturation and protein staining. The elastinolytic capacity of P. aeruginosa is reflected by clearing zones on nutrient agar plates containing insoluble elastin instead of casein. Mueller-Hinton agar plates on which S. aureus cells are grown as a lawn are used to assess the susceptibility of S. aureus isolates to staphylolysin. A clear zone around a staphylolysin-containing sample indicates inhibition of S. aureus growth. Methods for measuring the activity of individual proteases are based on their cleavage specificity. These include assays of elastinolytic activity of pseudolysin and/or staphylolysin using elastin-Congo red as a substrate, a method for determination of staphylolytic activity in which the rate of S. aureus cell lysis is determined spectrophotometrically, and methods for determination of peptidase activity of pseudolysin, staphylolysin, lysyl endopeptidase, and the aminopeptidase. The latter methods employ chromogenic or fluorogenic peptide derivatives comprising a short amino acid sequence matching the preferred cleavage site of the protease as substrates. As only one peptide bond is cleaved in each substrate, these assays permit kinetic studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Protease aus Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder
Sigma-Aldrich
Lysostaphin aus Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Aminopeptidase I from Streptomyces griseus, lyophilized powder, ≥200 units/mg protein
Sigma-Aldrich
Metalloendopeptidase Substrate