Merck
RfaH May Oppose Silencing by H-NS and YmoA Proteins during Transcription Elongation.
Journal of bacteriology (2022-03-09)
Bing Wang, Maura Mittermeier, Irina Artsimovitch
ZUSAMMENFASSUNG
Nucleoid-associated proteins (NAPs) silence xenogenes by blocking RNA polymerase binding to promoters and hindering transcript elongation. In Escherichia coli, H-NS and its homolog SptA interact with YmoA proteins Hha and YdgT to assemble nucleoprotein filaments that facilitate transcription termination by Rho, which acts in synergy with NusG. Countersilencing during initiation is facilitated by proteins that exclude NAPs from promoter regions, but auxiliary factors that alleviate silencing during elongation are not known. A specialized NusG paralog, RfaH, activates lipopolysaccharide core biosynthesis operons, enabling survival in the presence of detergents and antibiotics. RfaH strongly inhibits Rho-dependent termination by reducing RNA polymerase pausing, promoting translation, and competing with NusG. We hypothesize that RfaH also acts as a countersilencer of NAP/YmoA filaments. We show that deletions of hns and hha+ydgT suppress the growth defects of ΔrfaH by alleviating Rho-mediated polarity within the waa operon. The absence of YmoA proteins exacerbates cellular defects caused by reduced Rho levels or Rho inhibition by bicyclomycin but has negligible effects at a strong model Rho-dependent terminator. Our findings that the distribution of Hha and RfaH homologs is strongly correlated supports a model in which they comprise a silencing/countersilencing pair that controls expression of chromosomal and plasmid-encoded xenogenes. IMPORTANCE Horizontally acquired DNA drives bacterial evolution, but its unregulated expression may harm the recipient. Xenogeneic silencers recognize foreign genes and inhibit their transcription. However, some xenogenes, such as those encoding lipo- and exopolysaccharides, confer resistance to antibiotics, bile salts, and detergents, necessitating the existence of countersilencing fitness mechanisms. Here, we present evidence that Escherichia coli antiterminator RfaH alleviates silencing of the chromosomal waa operon and propose that plasmid-encoded RfaH homologs promote dissemination of antibiotic resistance genes through conjugation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DNAse I, Amplification Grade