Merck
Derivation of cerebellar neurons from human pluripotent stem cells.
Current protocols in stem cell biology (2012-03-15)
Slaven Erceg, Dunja Lukovic, Victoria Moreno-Manzano, Miodrag Stojkovic, Shomi S Bhattacharya
ZUSAMMENFASSUNG
Here we provide a protocol for differentiation of human embryonic stem cells (hESC) into cerebellar neurons using a novel defined culture method. This protocol is based on the application of inductive signaling factors involved in the early patterning of the cerebellar region of the neural tube, followed by the application of factors responsible for cerebellar neuron specification. Human pluripotent stem cells are induced to form spherical embryonic-like structures called embryoid bodies (EBs) and neuroepithelial tube-like rosettes using defined chemical conditions. In the presence of FGF, Wnt, and RA signaling factors the rosettes were specified to OTX2-expressing cells. Further specification of derived cells involves application of BMP factors involved in early development of granule cell progenitors, followed by mitogens and neurotrophins. It typically takes 5 weeks to generate the functional cerebellar granule neurons. This protocol is feeder-free, applies human recombinant factors, and produces high yield of desired neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus menschlicher Placenta, liquid, BioReagent, suitable for cell culture