Merck
Manganese Stress Tolerance Depends on Yap1 and Stress-Activated MAP Kinases.
International journal of molecular sciences (2022-12-24)
Inés G de Oya, Elena Jiménez-Gutiérrez, Hélène Gaillard, María Molina, Humberto Martín, Ralf Erik Wellinger
ZUSAMMENFASSUNG
Understanding which intracellular signaling pathways are activated by manganese stress is crucial to decipher how metal overload compromise cellular integrity. Here, we unveil a role for oxidative and cell wall stress signaling in the response to manganese stress in yeast. We find that the oxidative stress transcription factor Yap1 protects cells against manganese toxicity. Conversely, extracellular manganese addition causes a rapid decay in Yap1 protein levels. In addition, manganese stress activates the MAPKs Hog1 and Slt2 (Mpk1) and leads to an up-regulation of the Slt2 downstream transcription factor target Rlm1. Importantly, Yap1 and Slt2 are both required to protect cells from oxidative stress in mutants impaired in manganese detoxification. Under such circumstances, Slt2 activation is enhanced upon Yap1 depletion suggesting an interplay between different stress signaling nodes to optimize cellular stress responses and manganese tolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Glucose-6-Phosphate Dehydrogenase (G-6-PDH) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder