Merck
  • Intermittent compressive force regulates dentin matrix protein 1 expression in human periodontal ligament stem cells.
Intermittent compressive force regulates dentin matrix protein 1 expression in human periodontal ligament stem cells.
Journal of dental sciences (2023-01-17)
Jeeranan Manokawinchoke, Suconta Chareonvit, Vorapat Trachoo, Phoonsuk Limraksasin, Hiroshi Egusa, Thanaphum Osathanon
ZUSAMMENFASSUNG
Mechanical force differentially regulates periodontal ligament functions depending on types, magnitudes, and duration of stimulation. Intermittent compressive force (ICF) promotes an in vitro mineralization in human periodontal ligament cells. The present study investigated the effect of ICF on dentin matrix protein-1 (DMP1) expression in human periodontal ligament stem cells (hPDLSCs). Cells were treated with ICF in a serum-free culture medium for 24 h The mRNA and protein expression were examined using real-time polymerase chain reaction, immunofluorescence staining and Western blot analysis, respectively. The exposure to ICF in a serum-free condition significantly induced DMP1 expression in both mRNA and protein levels. The effect of ICF-induced DMP1 expression was inhibited by pretreatment with cycloheximide, indicating the requirement of the intermediated molecule(s). Pretreatment with transforming growth factor β (TGF-β) receptor inhibitor (SB431542) or neutralized antibody against TGF-β1 prior to ICF application abolished the effect of ICF-induced DMP1 expression. Further, recombinant TGF-β1 treatment stimulated DMP1 expression. The present study illustrated that ICF induces DMP1 expression in hPDLSCs via the regulation of TGF-β signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SB 431542 Hydrat, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Streptavidin−FITC from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥5 units/mg protein