Merck
  • Crosstalk between PML and p53 in response to TGF-β1: A new mechanism of cardiac fibroblast activation.
Crosstalk between PML and p53 in response to TGF-β1: A new mechanism of cardiac fibroblast activation.
International journal of biological sciences (2023-02-14)
Di Huang, Dan Zhao, Ming Li, Si-Yu Chang, Ya-Dong Xue, Ning Xu, Si-Jia Li, Nan-Nan Tang, Li-Ling Gong, Yi-Ning Liu, Hang Yu, Qing-Sui Li, Peng-Yu Li, Jia-Li Liu, Hai-Xin Chen, Ming-Bin Liu, Wan-Yu Zhang, Xing-Miao Zhao, Xian-Zhi Lang, Zhen-Dong Li, Yu Liu, Zhi-Yong Ma, Jia-Min Li, Ning Wang, Hai Tian, Ben-Zhi Cai
ZUSAMMENFASSUNG
Cardiac fibrosis is a common pathological cardiac remodeling in a variety of heart diseases, characterized by the activation of cardiac fibroblasts. Our previous study uncovered that promyelocytic leukemia protein (PML)-associated SUMO processes is a new regulator of cardiac hypertrophy and heart failure. The present study aimed to explore the role of PML in cardiac fibroblasts activation. Here we found that PML is significantly upregulated in cardiac fibrotic tissue and activated cardiac fibroblasts treated with transforming growth factor-β1 (TGF-β1). Gain- and loss-of-function experiments showed that PML impacted cardiac fibroblasts activation after TGF-β1 treatment. Further study demonstrated that p53 acts as the transcriptional regulator of PML, and participated in TGF-β1 induced the increase of PML expression and PML nuclear bodies (PML-NBs) formation. Knockdown or pharmacological inhibition of p53 produced inhibitory effects on the activation of cardiac fibroblasts. We further found that PML also may stabilize p53 through inhibiting its ubiquitin-mediated proteasomal degradation in cardiac fibroblasts. Collectively, this study suggests that PML crosstalk with p53 regulates cardiac fibroblasts activation, which provides a novel therapeutic strategy for cardiac fibrosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid