- The mechanism underlying redundant functions of the YTHDF proteins.
The mechanism underlying redundant functions of the YTHDF proteins.
Genome biology (2023-01-25)
Zhongyu Zou, Caraline Sepich-Poore, Xiaoming Zhou, Jiangbo Wei, Chuan He
PMID36694229
ZUSAMMENFASSUNG
The YTH N6-methyladenosine RNA binding proteins (YTHDFs) mediate the functional effects of N6-methyladenosine (m6A) on RNA. Recently, a report proposed that all YTHDFs work redundantly to facilitate RNA decay, raising questions about the exact functions of individual YTHDFs, especially YTHDF1 and YTHDF2. We show that YTHDF1 and YTHDF2 differ in their low-complexity domains (LCDs) and exhibit different behaviors in condensate formation and subsequent physiological functions. Biologically, we also find that the global stabilization of RNA after depletion of all YTHDFs is driven by increased P-body formation and is not strictly m6A dependent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete™, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%