Merck
  • Uptake and time-dependent subcellular localization of native and micellar curcumin in intestinal cells.
Uptake and time-dependent subcellular localization of native and micellar curcumin in intestinal cells.
BioFactors (Oxford, England) (2022-02-17)
Sandra Flory, Ann-Kathrin Benz, Jan Frank
ZUSAMMENFASSUNG
Uptake into intestinal cells and intracellular distribution into metabolically competent organelles, such as the endoplasmic reticulum, are important processes potentially limiting the bioavailability of xenobiotics. The incorporation of curcumin into polysorbate 80 micelles improves its naturally low oral bioavailability in humans. Here, we investigated uptake and time-dependent localization of curcumin in intestinal cells when administered as native or micellar formulation. Differentiated Caco-2 cells were incubated with 200 μmol/L native or micellar curcumin for up to 180 min and cellular uptake was quantified. Intracellular curcumin was detected already after 30 min and did not differ significantly between formulations or over time. Subcellular localization of native and micellar curcumin in Caco-2 cells was studied by density gradient centrifugation. After 30 min, curcumin from both formulations was mainly associated with mitochondria and lysosomes, after 180 min native curcumin was associated with mitochondria and peroxisomes, micellar curcumin with peroxisomes only. Uptake and localization of native and micellar curcumin in intestinal cells do not differ significantly and consequently do not explain differences in bioavailability in humans. The temporary co-localization with lysosomes is in agreement with the previously proposed role of endocytosis in cellular uptake of curcumin and warrants further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwer- und leichtkettenspezifisches Kaninchen-Anti- Kaninchen-IgG-Antikörper-Peroxidase-Konjugat, liquid, Calbiochem®