Merck
Assessment of stromal SCD-induced drug resistance of PDAC using 3D-printed zPDX model chips.
iScience (2023-01-03)
Chuntao Wu, Beiyuan Hu, Lei Wang, Xia Wu, Haitao Gu, Hanguang Dong, Jiuliang Yan, Zihao Qi, Qi Zhang, Huan Chen, Bo Yu, Sheng Hu, Yu Qian, Shuang Dong, Qiang Li, Xu Wang, Jiang Long
ZUSAMMENFASSUNG
Lipid metabolism is extensively reprogrammed in pancreatic ductal adenocarcinoma (PDAC). Stearoyl-coenzyme A desaturase (SCD) is a critical lipid regulator that was unexplored in PDAC. Here, we characterized the existence of cancer-associated fibroblasts (CAFs) with high SCD expression, and revealed them as an unfavorable prognostic factor. Therefore, primary CAFs and pancreatic cancer cells were harvested and genetically labeled. The mixture of CAFs and cancer cells were co-injected into scd-/-; prkdc-/-, or hIGF1/INS-expressing zebrafish to generate patient-derived xenograft models (zPDX). The models were aligned in 3D-printed chips for semi-automatic drug administration and high-throughput scanning. The results showed that chaperoning of the SCD-high CAFs significantly improved the drug resistance of pancreatic cancer cells against gemcitabine and cisplatin, while the administration of SCD inhibitors neutralized the protective effect. Our studies revealed the prognostic and therapeutic value of stromal SCD in PDAC, and proposed the application of zPDX model chips for drug testing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Insulin -Lösung human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Hyaluronidase aus Rindertestes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Anti-aktives β-Catenin(Anti-ABC)-Antikörper, Klon 8E7, clone 8E7, Upstate®, from mouse