- Transient Expression of Proteins Using COS Cells: Protein Expression.
Transient Expression of Proteins Using COS Cells: Protein Expression.
Current protocols in molecular biology (1991-04-01)
Alejandro Aruffo
PMID34266182
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroquin -diphosphat (Salz), powder or crystals, 98.5-101.0% (EP), meets EP testing specifications