Merck
  • Interstitial Macrophages Lead Early Stages of Bleomycin-Induced Lung Fibrosis and Induce Fibroblasts Activation.
Interstitial Macrophages Lead Early Stages of Bleomycin-Induced Lung Fibrosis and Induce Fibroblasts Activation.
Cells (2023-02-12)
Sofia Libório-Ramos, Catarina Barbosa-Matos, Raquel Fernandes, Caroline Borges-Pereira, Sandra Costa
ZUSAMMENFASSUNG
A progressive fibrosing phenotype is critical in several lung diseases. It is irreversible and associated with early patient mortality. Growing evidence has revealed pulmonary macrophages' role as modulators of the fibrotic processes. The proportion, phenotype, and function of alveolar (AM) and interstitial macrophages (IM) at the early stages of bleomycin-induced pulmonary fibrosis have not been clearly described. In this way, our study aimed to characterize these macrophage populations and investigate the effect on fibroblast activation. C57BL/6 mice were intratracheally injected with bleomycin and were sacrificed at day 3, 5, and 7 for the performance of flow cytometry and fluorescent-activated cell sorting analysis for protein and gene expression quantification. After bleomycin administration, the proportion of IM was significantly higher than that of AM, which showed a decay during the inflammatory phase, and peaked at day 7. At day 7 of the inflammatory phase, AM started shifting their phenotype from M1-like towards M2, while IM showed a M2-like phenotype. Conditioned medium derived from IM sorted at day 7 induced fibroblast activation and differentiation in myofibroblasts in vitro. Our findings indicate that IM are the largest macrophage population at the early stages of experimental pulmonary fibrosis and are secreted mediators able to activate fibroblasts, pointing to macrophage modulation as a potential therapeutic strategy to restrain progressive fibrosing lung disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DNAse I, grade II, from bovine pancreas
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)