Merck
A network map of the gastrin signaling pathway.
Journal of cell communication and signaling (2014-03-04)
Yashwanth Subbannayya, Kumari Anuja, Jayshree Advani, Urmesh Kumar Ojha, Vishalakshi Nanjappa, Bijesh George, Avinash Sonawane, Rekha V Kumar, Girija Ramaswamy, Akhilesh Pandey, B L Somani, Rajesh Raju
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gastrin I, human, ≥95% (HPLC), powder