Merck
  • Kazinol U inhibits melanogenesis through the inhibition of tyrosinase-related proteins via AMP kinase activation.
Kazinol U inhibits melanogenesis through the inhibition of tyrosinase-related proteins via AMP kinase activation.
British journal of pharmacology (2018-12-24)
Jihyun Lim, Sorim Nam, Ji Hye Jeong, Min Jung Kim, Young Yang, Myeong-Sok Lee, Hee Gu Lee, Jae-Ha Ryu, Jong-Seok Lim
ZUSAMMENFASSUNG
Kazinol U is a prenylated flavan isolated from an extract of Broussonetia kazinoki Sieb (Moraceae). Kazinol U has shown cytoprotective effects against cytokine-induced apoptotic cell death and induces AMP kinase (AMPK) activation through LKB1 activation. However, kazinol U has not been tested as a regulator of melanogenesis, although bark extract of B. kazinoki has been used as a cosmetic ingredient for skin conditioning. We cultured mouse, human melanoma cells and normal human melanocytes to demonstrate anti-melanogenic effects of kazinol U. A tyrosinase activity assay, Western blot, RT-qPCR and a luciferase reporter gene assay were performed to determine the anti-melanogenic mechanisms of kazinol U. We confirmed its effect on melanogenesis in vivo using zebrafish. Kazinol U inhibited the expression and activity of tyrosinase, the rate-limiting enzyme in melanogenesis, and reduced tyrosinase expression and activity in response to cAMP-inducing agents. Kazinol U reduced the expression of other melanogenic enzymes, such as tyrosinase-related protein (Tyrp) 1 and Tyrp2, and down-regulated microphthalmia-associated transcription factor (MITF), the master regulator of the tyrosinase gene family. Moreover, kazinol U induced phosphorylation of AMPK and MAPK proteins, which are MITF inhibitors. It also exhibited anti-melanogenic effects in zebrafish, a recently developed in vivo model. Our findings suggest that kazinol U reduces melanogenesis via its inhibitory effect on MITF and its downstream target genes, tyrosinase, Tyrp1 and Tyrp2. This work may provide a basis for the application of kazinol U for the treatment of hyperpigmentation skin disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Kazinol U, ≥95% (HPLC)