Merck
  • Dynamics of Endogenous Auxin and Its Role in Somatic Embryogenesis Induction and Progression in Cork Oak.
Dynamics of Endogenous Auxin and Its Role in Somatic Embryogenesis Induction and Progression in Cork Oak.
Plants (Basel, Switzerland) (2023-04-14)
Elena Carneros, Jorge Sánchez-Muñoz, Yolanda Pérez-Pérez, Beatriz Pintos, Aránzazu Gómez-Garay, Pilar S Testillano
ZUSAMMENFASSUNG
Somatic embryogenesis (SE) is a feasible in vitro regeneration system with biotechnological applications in breeding programs, although, in many forest species, SE is highly inefficient, mainly due to their recalcitrance. On the other hand, SE represents a valuable model system for studies on cell reprogramming, totipotency acquisition, and embryogenic development. The molecular mechanisms that govern the transition of plant somatic cells to embryogenic cells are largely unknown. There is increasing evidence that auxins mediate this transition and play a key role in somatic embryo development, although data on woody species are very limited. In this study, we analyzed the dynamics and possible role of endogenous auxin during SE in cork oak (Quercus suber L.). The auxin content was low in somatic cells before cell reprogramming, while it increased after induction of embryogenesis, as revealed by immunofluorescence assays. Cellular accumulation of endogenous auxin was also detected at the later stages of somatic embryo development. These changes in auxin levels correlated with the expression patterns of the auxin biosynthesis (QsTAR2) and signaling (QsARF5) genes, which were upregulated after SE induction. Treatments with the inhibitor of auxin biosynthesis, kynurenine, reduced the proliferation of proembryogenic masses and impaired further embryo development. QsTAR2 and QsARF5 were downregulated after kynurenine treatment. Our findings indicate a key role of endogenous auxin biosynthesis and signaling in SE induction and multiplication, as well as somatic embryo development of cork oak.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Auxin antibody, Mouse monoclonal, clone 1E11-C11, purified from hybridoma cell culture