- Magnetic Separation of Cell-Secreted Vesicles with Tailored Magnetic Particles and Downstream Applications.
Magnetic Separation of Cell-Secreted Vesicles with Tailored Magnetic Particles and Downstream Applications.
Methods in Molecular Biology (2023)
Bernuz, et al.
PMID37140802
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-CD81 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution